CLS55640500
Pyrex® class A low-actinic volumetric flask, Pyrex® ST stopper
capacity 500 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Joint
joint stopper19
feature
class A
packaging
case of 1 ea
manufacturer/tradename
Corning 55640-500
Corning 55640
tol. measuring range
0.20 mL
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
General description
Low actinic stained glass provides protection for materials sensitive to light. The protective color is an integral part of flask, which retains mechanical strength, chemical stability, and thermal resistance of Pyrex brand labware.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service