Key Documents
CLS4960250
Pyrex® round-bottom three neck vertical unequal height flask
capacity 250 mL, center joint: ST/NS 24/40
Synonym(s):
bottom flask, flask round, flask round bottom
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom flask
three-neck (Vertical)
Joint
center joint: ST/NS24/40
side joint ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper joint
packaging
pack of 1
case of 4
manufacturer/tradename
Corning 4960-250
capacity
250 mL
neck size
(3)
General description
With full length joints.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)