CLS3160102
Pyrex® petri dishes
complete, O.D. × H 100 mm × 20 mm
Synonym(s):
Corning® 3160
All Photos(1)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.43
Recommended Products
material
borosilicate glass
sterility
non-sterile
packaging
pack of 12
manufacturer/tradename
Corning 3160-102
O.D. × H
100 mm × 20 mm
Related Categories
General description
- PYREX® culture dishes are made of clear borosilicate glass.
- They can withstand repeated sterilization (wet or dry) and are chemically resistant.
- Beaded edges provide greater mechanical strength and reduce capillary action of condensed moisture.
- Top marked in blue enamel, bottom in white, enables easier sorting.
- Bottoms include a triangular, enamel reference point for serial dilutions.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
Meera Punjiya et al.
Biosensors & bioelectronics, 150, 111931-111931 (2020-01-14)
Transfection is a key function for many single-cell analyses. Reversible electroporation (EP) using high intensity electric fields is a simple means of transfection applicable to most cell types. For reversible EP, precise control over the electric field is critical to