Skip to Content
Merck
All Photos(1)

Key Documents

CF0044

Sigma-Aldrich

Tetrafluoropropyl azide

0.5 M in 1,2-dimethoxyethane

Synonym(s):

2,2,3,3-tetrafluoropropanyl azide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3F4N3
CAS Number:
Molecular Weight:
157.07
UNSPSC Code:
12352125
NACRES:
NA.06

Quality Level

concentration

0.5 M in 1,2-dimethoxyethane

SMILES string

FC(F)C(CN=[N+]=[N-])(F)F

InChI

1S/C3H3F4N3/c4-2(5)3(6,7)1-9-10-8/h2H,1H2

InChI key

CDMWKAMPYXSCJY-UHFFFAOYSA-N

General description

Tetrafluoropropyl azide is a fluoroalkyl azide reagent active in copper-catalyzed azide-alkyne cycloaddition (click reaction). The usage of this and other perfluoroalkyl reagents in this family allows for the previously challenging inclusion of fluorinated species as building blocks for click reactions.

Learn more about fluorinated click chemistry

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service