All Photos(1)
About This Item
Linear Formula:
C9H4F5IO2
CAS Number:
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22
Recommended Products
form
solid
reaction suitability
reaction type: C-C Bond Formation
storage temp.
−20°C
SMILES string
[I]1(OC(=O)c2c1cccc2)C(F)(F)C(F)(F)F
InChI
1S/C9H4F5IO2/c10-8(11,12)9(13,14)15-6-4-2-1-3-5(6)7(16)17-15/h1-4H
InChI key
KECFAALRJUVVHA-UHFFFAOYSA-N
Application
Togni-C2F5-reagent II is an extension of the existing Togni-CF3-reagent II (771147) showing exhibiting the same reactivity.
Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)
Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)
Legal Information
Product of CF Plus Chemicals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service