Skip to Content
Merck
All Photos(1)

Documents

CDS000492

Sigma-Aldrich

2-Hydroxybutanoic acid

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O3
CAS Number:
Molecular Weight:
104.10
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

form

solid

SMILES string

CCC(O)C(O)=O

InChI

1S/C4H8O3/c1-2-3(5)4(6)7/h3,5H,2H2,1H3,(H,6,7)

InChI key

AFENDNXGAFYKQO-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zulfiqar Ali Raza et al.
Bioprocess and biosystems engineering, 42(6), 901-919 (2019-02-28)
Polyhydroxyalkanoates (PHAs) are biological plastics that are sustainable alternative to synthetic ones. Numerous microorganisms have been identified as PHAs producers. They store PHAs as cellular inclusions to use as an energy source backup. They can be produced in shake flasks
Frank M Hauser et al.
Drug testing and analysis, 10(9), 1368-1382 (2018-04-19)
Chemical waste from the clandestine production of amphetamine is of forensic and environmental importance due to its illegal nature which often leads to dumping into the environment. In this study, 27 aqueous amphetamine waste samples from controlled Leuckart reactions performed
Covadonga Palacio et al.
Medicine, science, and the law, 61(1_suppl), 96-104 (2020-06-27)
Post-mortem changes of ions in the body fluids have been proposed as an objective tool for inferring the time of death. In particular, the post-mortem increase of potassium concentrations in the vitreous humour has gained great attention in the literature.
Xi-Yan Gao et al.
Applied microbiology and biotechnology, 104(7), 3183-3192 (2020-02-15)
Nitrate accumulation causes long-time threat to aquatic animals in recirculating aquaculture system (RAS); thus, nitrate removal is also required in RASs. However, the lack of carbon sources makes denitrification difficult to function. Nitrate removal performance of an aerobic denitrifying and
Miranda G M Kok et al.
Talanta, 197, 49-58 (2019-02-18)
Volumetric absorptive microsampling (VAMS) enables the collection of small and accurate quantities of biological fluids. Therefore, this sampling technique is of great interest for volume-limited samples or serial collection of samples. In this study, we examined the potential of VAMS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service