Key Documents
CAD6017
3-way stopcock
female Luer lock to male Luer lock, left side, female Luer lock
About This Item
UNSPSC Code:
41122000
NACRES:
NB.03
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science 6017
General description
The spring clip design holdsthe key in position under proper sealing pressure for optimum performance. There is no need for a screwdriver in cleaning operations.
  • Made from nickel plated brass or stainless steel
  • Tested to 45 psi water pressure
  • High pressure stopcocks are rated to 290 psi
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Can you autoclave this 3-way port? Can you use it to hook up on a sterile fermentation circuit?
    1 answer
    1. This product is supplied by Cadence Science. The manufacturer has not determined autoclavability for non-medical devices, such as this item. In general, chrome plated brass may be autoclaved, however brass is subject to corrosion. Other sources suggest an extended dry time when autoclaving. The suitability for use for fermentation systems would have to be determined experimentally by the end user.
      Helpful?