C112003
Cyclopentanecarboxylic acid
99%
Synonym(s):
Cyclopentanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C5H9CO2H
CAS Number:
Molecular Weight:
114.14
Beilstein:
2039729
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.453 (lit.)
bp
216 °C (lit.)
mp
3-5 °C (lit.)
density
1.053 g/mL at 25 °C (lit.)
SMILES string
OC(=O)C1CCCC1
InChI
1S/C6H10O2/c7-6(8)5-3-1-2-4-5/h5H,1-4H2,(H,7,8)
InChI key
JBDSSBMEKXHSJF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
200.1 °F - closed cup
Flash Point(C)
93.4 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Juan Manuel Gutierrez-Villagomez et al.
Talanta, 162, 440-452 (2016-11-14)
In this study, we report for the first time the efficiency of pentafluorobenzyl bromide (PFBBr) for naphthenic acid (NA) mixtures derivatization, and the comparison in the optimal conditions to the most common NAs derivatization reagents, BF
Naoto Tsuda et al.
PloS one, 9(11), e112027-e112027 (2014-11-19)
Diacylglycerol O-acyltransferase 1 (DGAT1) catalyzes the final committed step in triglyceride biosynthesis. DGAT1 null mice are known to be resistant to diet-induced obesity, and more insulin sensitive relative to the wild-type; however, the mice exhibit abnormalities in the skin. This
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service