All Photos(1)
About This Item
Linear Formula:
C6H11NCS
CAS Number:
Molecular Weight:
141.23
Beilstein:
774525
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.536 (lit.)
bp
219 °C (lit.)
density
0.996 g/mL at 25 °C (lit.)
SMILES string
S=C=NC1CCCCC1
InChI
1S/C7H11NS/c9-6-8-7-4-2-1-3-5-7/h7H,1-5H2
InChI key
MZSJGCPBOVTKHR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
203.0 °F - closed cup
Flash Point(C)
95 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Humaira Arshad et al.
Scientific reports, 10(1), 20439-20439 (2020-11-26)
This paper reports the enhanced performance of cholesterol oxidase (COx) conjugated CdSe/ZnS quantum dots (QDs) by using water-soluble mercaptoacitic acid (MAA) as linker. The functionalized MAA-CdSe/ZnS QDs conjugated in four different dilutions of cholesterol oxidase significantly affected QDs photoluminescence intensities
Peter Ulrich et al.
The American journal of tropical medicine and hygiene, 91(4), 824-832 (2014-09-04)
Because of emerging resistance to existing drugs, new chemical classes of antimalarial drugs are urgently needed. We have rationally designed a library of compounds that were predicted to accumulate in the digestive vacuole and then decrystallize hemozoin by breaking the
Fazila Rizvi et al.
Scientific reports, 9(1), 6738-6738 (2019-05-03)
A library of thiosemicarbazide derivatives of isoniazid 3-27, was synthesized and evaluated for their anti-inflammatory and urease inhibition activities, by using in vitro bioassays. Among these compounds 9, 10, 12, 21, and 26 were identified as new derivatives. Prolonged use
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service