Key Documents
BAF247390003
SP Bel-Art® Poxygrid Glove Dispenser
epoxy-coated (wire), to hold, Triple Box Holder- holds 3x glove box
Synonym(s):
Glove box holder, glove dispenser
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
material
epoxy-coated (wire)
feature
to hold Triple Box Holder- holds 3x glove box
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F24739-0003
H × L × width
8.25 in. (210 mm) × 17.5 in. (432 mm) × 4.25 in. (108 mm)
General description
Keep disposable gloves handy with these welded steel wire racks. Select rack to hold one, two or three boxes of disposable gloves, wipes or tissues.
Features and Benefits
  • Glove box drops into an individual retaining compartment and lifts out easily for replacement
  • Retaining compartment internal dimensions are 12.7cmW x 9.5cmD (5 x 3¾")
  • Designed for mounting to a vertical surface
  • Durable, blue epoxy coated racks are steam autoclavable at 121°C (250°F)
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)