Skip to Content
Merck
All Photos(2)

Key Documents

B86206

Sigma-Aldrich

cis-2-Butene-1,4-diol

97%

Synonym(s):

(2Z)-2-Butene-1,4-diol, (Z)-1,4-Dihydroxy-2-butene, (Z)-2-Buten-1,4-diol, (Z)-2-Butene-1,4-diol, cis-1,4-Dihydroxy-2-butene, cis-But-2-en-1,4-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
1679241
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.478 (lit.)

bp

235 °C (lit.)

mp

4-10 °C (lit.)

density

1.072 g/mL at 20 °C (lit.)

SMILES string

[H]\C(CO)=C(/[H])CO

InChI

1S/C4H8O2/c5-3-1-2-4-6/h1-2,5-6H,3-4H2/b2-1-

InChI key

ORTVZLZNOYNASJ-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2

Target Organs

Liver,Kidney,Blood

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanke Liang et al.
The Journal of organic chemistry, 76(24), 9962-9974 (2011-10-28)
The first psico-oxetanocin analogue of the powerful antiviral natural product, oxetanocin A, has been readily synthesized from cis-2-butene-1,4-diol. Key 2-methyleneoxetane precursors were derived from β-lactones prepared by the carbonylation of epoxides. F(+)-mediated nucleobase incorporation provided the corresponding nucleosides in good
T Okuda et al.
The Journal of antibiotics, 37(7), 718-722 (1984-07-01)
A novel antibiotic, penitricin, Ro 09-0804, has been produced in the culture filtrate of Penicillium aculeatum NR 5165. This antibiotic was purified by repeated extraction of culture filtrate with 1-butanol, and passage of the crude extracts through Sephadex G-10, followed
Janusz Kozakiewicz et al.
Materials (Basel, Switzerland), 13(21) (2020-11-11)
The results of studies on the synthesis and characterization of conductive polymer composite materials designed as potential separators for lithium ion batteries are presented. The conductive polymer composites were prepared from UV-cured poly(siloxane-urethanes)s (PSURs) containing poly(ethylene oxide) (PEO) segments and
Matthew K Kiesewetter et al.
Journal of the American Chemical Society, 133(41), 16390-16393 (2011-09-24)
The catalytic condensation of cis-2-butene-1,4-diol with CpRu(MQA)(C(3)H(5)) (Cp = cyclopentadienyl, MQA = 4-methoxyquinoline-2-carboxylate) generates poly(2-butenediol), an unsaturated telechelic polyether diol with molecular weights between 400 and 4600 g/mol. This Ru(IV) allyl catalyst enchains 2-butene-1,4-diol primarily as the linear trans-2-butenyl ether
Yu-ying Zheng et al.
Guang pu xue yu guang pu fen xi = Guang pu, 25(3), 402-404 (2005-07-15)
The mechanism of the modification of Kevlar fiber by polypropylene glycol(PPG) and cis-2-butene-1, 4-diol was studied in the paper, the authors learned the esterification of toluene-2, 4-diisocyanate (TDI) onto Kevlar fiber by infrared spectrum. In the mean time, the infrared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service