Skip to Content
Merck
All Photos(1)

Documents

B73104

Sigma-Aldrich

1-Bromonaphthalene

97%

Synonym(s):

1-Naphthyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7Br
CAS Number:
Molecular Weight:
207.07
Beilstein:
1906414
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.6570 (lit.)

bp

133-134 °C/10 mmHg (lit.)

mp

−2-−1 °C (lit.)

density

1.48 g/mL at 20 °C (lit.)

SMILES string

Brc1cccc2ccccc12

InChI

1S/C10H7Br/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H

InChI key

DLKQHBOKULLWDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromonaphthalene serves as an intermediate for the synthesis of biaryl compounds via cross-coupling reactions.

Application

1-Bromonaphthalene is a bromoarene that can be used in:
  • Palladium-catalyzed Suzuki–Miyaura coupling reaction with potassium aryltrifluoroborates without the use of phase-transfer catalysts or phosphine ligands.
  • The preparation of indeno annelated polycyclic aromatic hydrocarbons by reacting with o-bromobenzeneboronic acid and oligocyclic bromoarenes via Suzuki-Heck type coupling.
  • Ni catalyzed Kumada–Tamao–Corriu cross-coupling reaction with PhMgBr.
  • The preparation of arylnaphthalenes via palladium-catalyzed Suzuki-Miyaura cross-coupling reaction with aryl boronic acid.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exploring the selectivity of the Suzuki-Miyaura cross-coupling reaction in the synthesis of arylnaphthalenes
Lima CFRAC, et al.
Tetrahedron, 67(4), 689-697 (2011)
High efficiency of cavity-based triaryl-phosphines in nickel-catalysed Kumada-Tamao-Corriu cross-coupling
Monnereau L, et al.
Chemical Communications (Cambridge, England), 47(23), 6626-6628 (2011)
Xinzhen Du et al.
Journal of colloid and interface science, 274(2), 645-651 (2004-05-18)
The binding of polyethylene glycol (10) n-octylphenyl ether (OPE) and polyethylene glycol (10) tert-octylphenyl ether (Triton X-100, TX) to beta-cyclodextrin (beta-CD) and heptakis(2,3- beta-dimethyl)- beta-CD (DM- beta-CD) was described in detail by surface tension, steady-state fluorescence of OPE and TX
A New Suzuki- Heck-Type Coupling Cascade: Indeno [1, 2, 3]-Annelation of Polycyclic Aromatic Hydrocarbons
Wegner HA, et al.
The Journal of Organic Chemistry, 68(3), 883-887 (2003)
Xin-zhen Du et al.
Guang pu xue yu guang pu fen xi = Guang pu, 24(4), 402-406 (2005-03-16)
Room-temperature phosphorescence of 1-BrN induced by a combination of OPE-10 and Triton X-100 with beta-CD was comparatively studied. In terms of molecular size and dimensions of beta-CD, the octyl group and phenyl group of OPE-10 and Triton X-100 were incorporated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service