Skip to Content
Merck
All Photos(1)

Key Documents

ALD00508

Sigma-Aldrich

Yu Fluorination Ligand

≥95%

Synonym(s):

2,5,6,8-tetramethyl-3,4-dihydro-2H-pyrano[2,3-b]quinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19NO
CAS Number:
Molecular Weight:
241.33
MDL number:
UNSPSC Code:
41116155
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation

SMILES string

CC1=C2C(OC(C)CC2)=NC3=C1C(C)=CC(C)=C3

InChI

1S/C16H19NO/c1-9-7-10(2)15-12(4)13-6-5-11(3)18-16(13)17-14(15)8-9/h7-8,11H,5-6H2,1-4H3

InChI key

LOKRNFHHPZEJLL-UHFFFAOYSA-N

General description

Yu Fluorination Ligand is widely employed for the chemoselective functionalization of sp2- and sp3 C-H bonds.

Application

The Yu Fluorination Ligand in tandem with Selectfluor(R) (Aldrich 439479) under palladium catalysis can promote direct fluorination of C(sp3)-H bonds stereoselectively. Yu and coworkers have displayed this on amino acid derivatives with directing group assistance from the Yu-Wasa Auxiliary (Aldrich 791806).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service