A65409
3-Picolylamine
≥99%
Synonym(s):
3-(Aminomethyl)pyridine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
Beilstein:
108056
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥99%
form
liquid
refractive index
n20/D 1.551 (lit.)
bp
73-74 °C/1 mmHg (lit.)
mp
−21 °C (lit.)
density
1.062 g/mL at 25 °C (lit.)
SMILES string
NCc1cccnc1
InChI
1S/C6H8N2/c7-4-6-2-1-3-8-5-6/h1-3,5H,4,7H2
InChI key
HDOUGSFASVGDCS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
212.0 °F
Flash Point(C)
100 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A Kessentini et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 191, 241-248 (2017-10-19)
The current work undertakes the growth and the physicochemical properties of a novel green-yellow luminescence semi-organic material, the 3-picolylammonium bromide abbreviated (Pico-Br). In this paper, we report the X-ray diffraction measurements which show that the crystal lattice consists of distinct
Allergic contact dermatitis from nicotinic acid esters.
M Audicana et al.
Contact dermatitis, 22(1), 60-61 (1990-01-01)
Jian-yu Guo et al.
Guang pu xue yu guang pu fen xi = Guang pu, 26(3), 460-462 (2006-07-13)
Raman spectra of human serum albumin (HSA) and HSA-3-picolylamine complex were obtained. The spectra indicate the configuration and structural transformations of HSA. The results show that the secondary structure is main alpha-helix, and the binding of 3-picolylamine doesn't change the
Ligand oxidation of a deprotonated bis(picolyl)amine Ir(I)(cod) complex.
Cristina Tejel et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(35), 10932-10936 (2008-11-20)
John Williams et al.
Bioanalysis, 12(3), 143-158 (2020-02-14)
Aim: Very long chain fatty acids (VLCFAs) have been identified as biomarkers for several peroxisomal disorders necessitating the need for reliable biomarker assays in particular C20, C22, C24, C26 in cerebrospinal fluid (CSF). Until now no absolute quantitation assay for
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service