Skip to Content
Merck
All Photos(1)

Key Documents

94565

Sigma-Aldrich

Pentyl valerate

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)2CH2COO(CH2)4CH3
CAS Number:
Molecular Weight:
172.26
Beilstein:
1754427
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.417

bp

201-203 °C (lit.)

density

0.865 g/mL at 20 °C (lit.)

functional group

ester

SMILES string

CCCCCOC(=O)CCCC

InChI

1S/C10H20O2/c1-3-5-7-9-12-10(11)8-6-4-2/h3-9H2,1-2H3

InChI key

FGPPDYNPZTUNIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tripti Raghavendra et al.
BioMed research international, 2014, 353845-353845 (2014-08-06)
Pentyl valerate was synthesized biocatalytically using Candida rugosa lipase (CRL) immobilized in microemulsion based organogels (MBGs). The optimum conditions were found to be pH 7.0, temperature of 37 °C, ratio of concentration of water to surfactant (Wo) of 60, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service