Skip to Content
Merck
All Photos(1)

Key Documents

936022

Sigma-Aldrich

Sodium hexafluorophosphate

greener alternative

99.9% trace metals basis, anhydrous, battery grade

Synonym(s):

NaPF6, Sodium hexafluorophosphate(v)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaPF6
CAS Number:
Molecular Weight:
167.95
MDL number:
UNSPSC Code:
12352104

grade

battery grade

Quality Level

Assay

99.9% trace metals basis

form

white powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤100 ppm Cl
≤100 ppm Sulfate
≤1500 ppm trace metals

mp

>200 °C (lit.)

density

2.369 g/mL at 25 °C (lit.)
2.369 g/mL

application(s)

battery manufacturing

greener alternative category

SMILES string

[Na+].F[P-](F)(F)(F)(F)F

InChI

1S/F6P.Na/c1-7(2,3,4,5)6;/q-1;+1

InChI key

KMADQUOCJBLXRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hexafluorophosphate is a white powder and is soluble in water and polar organic solvents, including many of the carbonates and ethers commonly used in liquid battery electrolytes. Our battery grade NaPF6 meets both high level of purity and low moisture to ensure reliable and consistent performance for battery research.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Battery grade sodium hexafluorophosphate (NaPF6) is a reliable precursor salt for sodium ion batteries (NIBs). NaPF6 is commonly used as the source of sodium ions in liquid electrolytes due to its excellent properties including high solubility in organic solvents, high anodic electrochemical stability, and ability to form fluoride-rich solid electrolyte interphases (SEI) Often NaPF6 is used in research to design electrolytes that enhance the energy/power density or cycle life of sodium-ion batteries. For example, studies have shown that NaPF6-based electrolytes often offer the best performance, especially in conjunction with additives like FEC Additionally, NaPF6 is widely used because it is compatible with most battery materials including sodium metal, hard carbon, most oxide and phosphate cathode materials, as well as copper and aluminum current collectors. As a result, NaPF6-based electrolytes are the go-to for sodium-ion battery research and development.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of the electrolyte salt anion on the solid electrolyte interphase formation in sodium ion batteries
Eshetu G G, et al.
Nano Energy, 55, 327-340 (2019)
Comparison of Ionic Transport Properties of Non-Aqueous Lithium and Sodium Hexafluorophosphate Electrolytes
Landesfeind J, et al.
Journal of the Electrochemical Society, 168 (2021)
SEI Composition on Hard Carbon in Na-Ion Batteries After Long Cycling: Influence of Salts (NaPF6, NaTFSI) and Additives (FEC, DMCF)
Fondard J, et al.
Journal of the Electrochemical Society, 167, 070526-070526 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service