Skip to Content
Merck
All Photos(2)

Key Documents

932760

Sigma-Aldrich

3,4,9,10-Perylenetetracarboxylic dianhydride

Synonym(s):

PTCDA, Perylenetetracarboxylic acid dianhydride, Perylene-3,4,9,10-tetracarboxylic dianhydride, Pigment Red 224

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H8O6
CAS Number:
Molecular Weight:
392.32
Beilstein:
57831
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.21

Assay

99% (sublimed)

Quality Level

mp

>300 °C

solubility

DMSO: slightly soluble

Orbital energy

HOMO 5.85 eV 
LUMO 3.9 eV 

SMILES string

O1C(=O)c2c3c4c(c5c6c(c4cc2)ccc7c6c(cc5)C(=O)OC7=O)ccc3C1=O

InChI

1S/C24H8O6/c25-21-13-5-1-9-10-2-6-15-20-16(24(28)30-23(15)27)8-4-12(18(10)20)11-3-7-14(22(26)29-21)19(13)17(9)11/h1-8H

InChI key

CLYVDMAATCIVBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,4,9,10-Perylenetetracarboxylic dianhydride, also known as Perylenetetracarboxylic acid dianhydride, is a large π-conjugated semiconductor aromatic organic molecule. It is an organic dye molecule and an organic semiconductor. It is used as a precursor to a class of molecules known as Rylene dyes, which are useful as pigments and dyes. It is a dark red solid with low solubility in aromatic solvents.
The compound has attracted much interest as an organic semiconductor. It has potential applications in organic and molecular electronics, and typically used as an archetype molecular compound. PTCDA shares many properties with conventional semiconductors (with their delocalized electronic states), and insulator-like organic molecular crystals (OMCs) (with their large absorption oscillator strengths, excitonic self-trapping, polaron-assisted conduction, etc.). That is due to extremely close π-π stacking, an unusually small intermolecular distance of only 3.21 A. Among its applications, it is important to highlight the latest focus on environment monitoring, self-assembly and photocatalysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenwu Zhou et al.
The Science of the total environment, 780, 146483-146483 (2021-03-28)
Environment pollution is getting serious and various poisonous contaminants with chemical durability, biotoxicity and bioaccumulation have been widespreadly discovered in municipal wastewaters and surface water. The detection and removal of pollutants show great significance for the protection of human health
Zijian Zhang et al.
Advanced materials (Deerfield Beach, Fla.), 32(32), e1907746-e1907746 (2020-07-01)
A highly crystalline perylene imide polymer (Urea-PDI) photocatalyst is successfully constructed. The Urea-PDI presents a wide spectrum response owing to its large conjugated system. The Urea-PDI performs so far highest oxygen evolution rate (3223.9 µmol g-1 h-1 ) without cocatalysts
Mohammad Rezwan Habib et al.
Nanoscale, 10(34), 16107-16115 (2018-08-17)
We report the photoluminescence (PL) characteristics of a van der Waals (vdW) heterojunction constructed by simply depositing an organic semiconductor of 3,4,9,10-perylene tetracarboxylic dianhydride (PTCDA) onto a two-dimensional MoS2 monolayer. The crystallinity of PTCDA on MoS2 is significantly improved due
Kangyi Kong et al.
Chemical communications (Cambridge, England), 55(56), 8090-8093 (2019-06-25)
In this communication, a self-assembled supramolecular system consisting of phosphoric acid substituted perylene diimide (P-PMPDI) has been successfully developed for highly efficient photocatalytic hydrogen evolution. Compared with a carboxylic substituent perylene diimide (P-CMPDI), P-PMPDI showed a superior H2 evolution reaction
Weiqin Wei et al.
Small (Weinheim an der Bergstrasse, Germany), 15(49), e1903933-e1903933 (2019-10-28)
A semi-core-shell structure of perylene diimide (PDI) self-assembly coated with TiO2 nanoparticles is constructed, in which nanoscale porous TiO2 shell is formed and PDI self-assembly presented 1D structure. A full-spectrum photocatalyst is obtained using this structure to resolve a conundrum-TiO2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service