Skip to Content
Merck
All Photos(1)

Key Documents

930911

Sigma-Aldrich

Praseodymium nitrate hexahydrate

99.99% (trace rare earth metals basis)

Synonym(s):

Praseodymium(III) nitrate hexahydrate, Nitric acid praseodymium(3+) salt, Praseodymium trinitrate, Praseodymium trinitrate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pr(NO3)3·6H2O
CAS Number:
Molecular Weight:
435.01
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.54
Assay:
99.99% (trace rare earth metals basis)
form:
crystals
solubility:
H2O: soluble ((lit.))

Quality Level

Assay

99.99% (trace rare earth metals basis)

form

crystals

impurities

≤150 ppmtrace (rare earth metals)
≤500 ppm (trace metals)

mp

<100 °C ((lit.))

solubility

H2O: soluble ((lit.))

density

2.233 g/cm3 ((lit.))

SMILES string

O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.6H2O.Pr/c3*2-1(3)4;;;;;;;/h;;;6*1H2;/q3*-1;;;;;;;+3

InChI key

LXXCECZPOWZKLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Praseodymium nitrate is a green, crystalline salt commonly found as a hexahydrate. The salt is soluble in polar solvents including water, alcohols, amines, ethers, and acetonitrile. Like many trivalent nitrates, praseodymium nitrate has a low thermal decomposition temperature with onset temperature of 100 °C. Praseodymium nitrate is produced from ores rich in rare earth metals. Dissolving the ores in nitric acid yields solutions of rare earth nitrates, which can be separated by solvent extraction, isolated into individual rare earth metals, and purified.

Application

Praseodymium nitrate is used in the preparation of praseodymium-doped materials and praseodymium compounds because it is one of the most convenient sources of praseodymium ions. Owing to its high solubility and low thermal decomposition temperature, praseodymium is a particularly useful reagent in reactions that use sol-gel processing and hydrothermal-calcination methods. Like other rare earth metals, praseodymium has a cloud of shielded f-electrons, which allow for long excited state lifetimes and high luminescence yields. Because of these properties, Pr-doped materials and Pr-compounds are used as phosphors in optics and added to oxides used in ceramics for colorful effects. Researchers continue to find ways to leverage the unique optical effects. For example, praseodymium metal-organic frameworks synthesized using Pr(NO3)3 have been explored for luminescence-sensing of small molecules and Pr-doped metal oxides researched for improved optical thermometry . Another common application of praseodymium nitrate is the synthesis of double perovskite oxides for solid-oxide fuel cells (SOFC). For example, researchers have shown that praseodymium cobaltites, praseodymium manganates, and praseodymium nickelates offer excellent ionic and electronic conductivities for SOFCs at intermediate temperatures above 400 °C. The unique band structure afforded by Pr3+ ions stabilizes oxygen vacancies, which are integral to ionic diffusion.

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Ox. Sol. 3 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-sustainable protonic ceramic electrochemical cells using a triple conducting electrode for hydrogen and power production.
Ding, H., et al.
Nature Communications, 11, 1907-1907 (2020)
Intermediate-temperature electrochemical performance of a polycrystalline PrBaCo2O5+delta cathode on samarium-doped ceria electrolyte
Chen, D., et al.
Journal of Power Sources, 188, 96-105 (2009)
Mechanistic insights into the phase transition and metal ex-solution phenomena of Pr0.5Ba0.5Mn0.85Co0.15O3?? from simple to layered perovskite under reducing conditions and enhanced catalytic activity.
Kim, K., et al.
Energy & Environmental Science, 14, 873-882 (2021)
A Novel Optical Thermometry Strategy Based on Diverse Thermal Response from Two Intervalence Charge Transfer States.
Gao, Y., et al.
Advances in Functional Materials, 26, 3139?3145-3139?3145 (2016)
Praseodymium and gadolinium doped ceria as a cathode material for low temperature solid oxide fuel cells.
Chockalingam, R., et al.
Journal of Power Sources, 250, 80-89 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service