Skip to Content
Merck
All Photos(1)

Key Documents

926248

Sigma-Aldrich

Redox Responsive Poly(ethylene glycol)-block-poly(lactide-alt-glycolide)

PEG average Mn 5000, PLGA Mn 15000

Synonym(s):

Drug release, PEG-PLGA with disulfide linkage, PEG-SS-PLGA, Stimuli-responsive

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(C2H4O)nC7H12O4S2(C5H6O4)m
UNSPSC Code:
12352112
NACRES:
NA.23

form

powder, chunks or granules

Quality Level

mol wt

PEG average Mn 5000
PLGA Mn 15000

storage temp.

2-8°C

SMILES string

O=C(C(C)OC(CO[H])=O)OCCSSCCOC(OCCOCCOC)=O

InChI

1S/C15H26O10S2/c1-12(25-13(17)11-16)14(18)22-7-9-26-27-10-8-24-15(19)23-6-5-21-4-3-20-2/h12,16H,3-11H2,1-2H3

InChI key

DPMVWUUTGXWXPV-UHFFFAOYSA-N

Application

PEG-SS-PLGA is a redox responsive, biocompatible, amphiphilic block copolymer composed of a hydrophilic PEG block and a hydrophobic poly(lactide-alternative-glycolide) (PLGA) block, with a reducible disulfide linkage between blocks. This polymer can be fabricated into Glutathione (GSH) responsive nanoparticles which can be used as an intracellular drug delivery system.

Different from common Poly(lactide-co-glycolide) (PLGA) polymer, the lactic and glycolic units in Poly(lactide-alt-glycolide) block alternate in sequence in the polymer chain, and the molar ratio of lactic to glycolic units in the polymer is exactly 1:1.

Poly(lactide-alt-glycolide) exhibits excellent solubility in common organic solvents, such as acetonitrile, acetone, dioxane, DCM, and THF, providing great convenience for drug delivery researches and applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Conte et al.
Journal of controlled release : official journal of the Controlled Release Society, 277, 126-141 (2018-03-15)
In the treatment of lung cancer, there is an urgent need of innovative medicines to optimize pharmacological responses of conventional chemotherapeutics while attenuating side effects. Here, we have exploited some relatively unexplored subtle differences in reduction potential, associated with cancer
Lin Yu et al.
Biomacromolecules, 12(4), 1290-1297 (2011-03-03)
This paper reports the influence of sequence structures of block copolymers composed of poly(lactic acid-co-glycolic acid) (PLGA) and poly(ethylene glycol) (PEG) on their thermogelling aqueous behaviors. A series of thermogelling PLGA-PEG-PLGA triblock copolymers with similar chemical compositions and block lengths

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service