Skip to Content
Merck
All Photos(1)

Documents

925713

Sigma-Aldrich

Platinum(II) 2,3,7,8,12,13,17,18-octaethyl-21H,23H-porphyrin

≥97%

Synonym(s):

Platinum octaethylporphyrin, Platinum(II) 2,3,7,8,12,13,17,18-octaethyl-21H,23H-porphyrin, PtOEP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H44N4Pt
CAS Number:
Molecular Weight:
727.84
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

UV: 381, 501, 535 nm (in CHCl3)

Quality Level

Assay

≥97%

form

powder or crystals

color

red

mp

>300 °C

λmax

649 nm (PL)

Orbital energy

HOMO 5.3 eV 
LUMO 3.2 eV 

UV absorption

λ: 381 nm Amax
λ: 501 nm Amax
λ: 535 nm Amax

SMILES string

CCc1c(CC)c2cc3c(CC)c(CC)c4cc5nc(cc6c(CC)c(CC)c(cc1n2)n6[Pt]n34)c(CC)c5CC

InChI

1S/C36H44N4.Pt/c1-9-21-22(10-2)30-18-32-25(13-5)26(14-6)34(39-32)20-36-28(16-8)27(15-7)35(40-36)19-33-24(12-4)23(11-3)31(38-33)17-29(21)37-30;/h17-20H,9-16H2,1-8H3;/q-2;+2/b29-17-,30-18-,31-17-,32-18-,33-19-,34-20-,35-19-,36-20-;

InChI key

WAODGUVBNLMTSF-XTPDIVBZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Platinum octaethylporphyrin (PtOEP) is used as a red triplet emitting material with high external quantum yields for organic light-emitting diodes (OLEDs). It can be utilized as the active component in chemical sensors or biosensors research. The ability of these compounds to form ordered structures makes them interesting candidates for molecular electronics, molecular magnets, and other advanced materials. 4) It is suitable to be used as a photosensitizer in photodynamic therapy.
Platinum octaethylporphyrin (PtOEP) may be used as a fluorescent dye in the fabrication of polymer- and silica-based oxygen sensors.
It is used as a red triplet emitting material with high external quantum yields for OLEDs. PtOEP is used as a dopant in host materials like CBP and Alq3. Its red emission is near 650 nm depending on host material and processing conditions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Prototype of a Volumetric Three-Dimensional Display Based on Programmable Photo-Activated Phosphorescence.
Dr Wan S, et al.
Angewandte Chemie (International Edition in English), 59, 8416-8420 (2020)
Highly efficient deep-red organic light-emitting diodes using exciplex-forming co-hosts and thermally activated delayed fluorescence sensitizers with extended lifetime.
Li D, et al.
Journal of Material Chemistry C, 7, 9531-9536 (2019)
Photoelectrochemical Properties of Metalloporphyrins.
Kampas F J, et al.
Nature, 284, 40-42 (1980)
Organic Electroluminescent Sensor for Pressure Measurement.
Matsuda Y, et al.
Sensors, 12, 13899-13906 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service