Skip to Content
Merck
All Photos(1)

Key Documents

925446

Sigma-Aldrich

5-Vinylthianthrenium tetrafluoroborate

≥95%

Synonym(s):

Vinyl-TT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11BF4S2
CAS Number:
Molecular Weight:
330.17
UNSPSC Code:
12352108
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

110-112 °C

storage temp.

−20°C

SMILES string

[F-][B+3]([F-])([F-])[F-].S1C=2C=CC=CC2[S+](C=C)C=3C=CC=CC13

InChI

1S/C14H11S2.BF4/c1-2-16-13-9-5-3-7-11(13)15-12-8-4-6-10-14(12)16;2-1(3,4)5/h2-10H,1H2;/q+1;-1

InChI key

FXHBWPNFHMRFKM-UHFFFAOYSA-N

Application

5-Vinylthianthrenium tetrafluoroborate is a crystalline, shelf-stable vinylating reagent developed by the Ritter Lab. Despite the great functionality of the vinyl group for diversification of target compounds, the selective installation of this moeity has remained challenging. 5-Vinylthianthrenium tetrafluoroborate can perform this otherwise challenging transformation with a broad scope of substrate, including the N-vinylation of heterocycles, vinylation of aryl boronic acids, and the annulation of cyclic and heterocyclic substrates. Additionally, this reagent can be used to enable palladium catalyzed cross-coupling reactions. The functionality of 5-vinylthianthrenium tetrafluoroborate enables the usage of vinyl groups as a versatile tool for further functionalization of many substrates and is a functional tool for many snythetic pathways.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemoselective umpolung of thiols to episulfoniums for cysteine bioconjugation
Hartmann P, et al.
Nature Chemistry, 16, 380-388 (2024)
Vinyl Thianthrenium Tetrafluoroborate: A Practical and Versatile Vinylating Reagent Made from Ethylene
Fabio Julia, et al
Journal of the American Chemical Society, 143, 33, 12992?12998-12992?12998 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service