Skip to Content
Merck
All Photos(2)

Key Documents

918970

Sigma-Aldrich

VincePhos

≥95%

Synonym(s):

7,7,9,9-Tetramethyl-8-(2′,4′,6′-triisopropyl-3,6-dimethoxy-[1,1′-biphenyl]-2-yl)-1,4-dioxa-8-phosphaspiro[4.5]decane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H51O4P
CAS Number:
Molecular Weight:
554.74
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

reagent type: ligand

functional group

phosphine

storage temp.

−20°C

SMILES string

P3(C(CC4(OCCO4)CC3(C)C)(C)C)c1c(ccc(c1c2c(cc(cc2C(C)C)C(C)C)C(C)C)OC)OC

InChI

1S/C34H51O4P/c1-21(2)24-17-25(22(3)4)29(26(18-24)23(5)6)30-27(35-11)13-14-28(36-12)31(30)39-32(7,8)19-34(20-33(39,9)10)37-15-16-38-34/h13-14,17-18,21-23H,15-16,19-20H2,1-12H3

InChI key

BYFHGEFWDTYSSA-UHFFFAOYSA-N

Application

VincePhos is a biaryl phosphorinane ligand for Pd catalysis. This ligand has been used in a variety of Pd-catalyzed reactions including the sulfonamidation of aryl halides, C-O couplings, and C-N couplings.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David M Barnes et al.
The Journal of organic chemistry, 84(8), 4873-4892 (2019-01-11)
Dasabuvir (1) is an HCV polymerase inhibitor which has been developed as a part of a three-component direct-acting antiviral combination therapy. During the course of the development of the synthetic route, two novel coupling reactions were developed. First, the copper-catalyzed
Pd-Catalyzed Cross-Coupling Reactions Promoted by Biaryl Phosphorinane Ligands.
Laffoon S D, et al.
ACS Catalysis, 9(12), 11691-11708 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service