Skip to Content
Merck
All Photos(1)

Key Documents

918962

Sigma-Aldrich

Poly(ethylene glycol) methyl ether-block-poly(D,L lactide)

PEG average Mn 5000, PDLA average Mn 50000

Synonym(s):

Biodegradable, Block copolymer, Drug delivery, PEG-PLA, PLLA, mPEG-PLA, mPEG-PLLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO[CH(CH3)COO]m[CH2CH2O]nCH3
UNSPSC Code:
12352106
NACRES:
NA.23

form

solid

Quality Level

mol wt

PDLA average Mn 50,000 (by NMR)
PDLA average Mn 50000
PEG average Mn 5,000 (by NMR)
PEG average Mn 5000

color

white to yellow

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Poly(ethylene glycol) methyl ether-block-poly(D,L lactide) is an amphiphilic, diblock copolymer composed of a hydrophilic PEG block and a hydrophobic PLA block. These biodegradable, biocompatible polymers can self-assemble to form nanoparticles, such as micelles and polymersomes, in both aqueous and non-aqueous media. Due to these properties, these polymers are widely used in polymeric nanoparticle formulation to achieve controlled and targeted delivery of therapeutic agents (e.g. APIs, genetic material, peptides, vaccines, and antibiotics).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cynthia Griggins et al.
HEC forum : an interdisciplinary journal on hospitals' ethical and legal issues, 23(1), 43-56 (2010-12-21)
This essay describes and critically evaluates a co-operative educational program to train Ugandan health care workers in bioethics. It describes one "bottom-up" effort, a week-long intensive workshop in bioethics provided by the authors to health care professionals in a developing
Ren Zhong Xiao et al.
International journal of nanomedicine, 5, 1057-1065 (2010-12-21)
Due to their small particle size and large and modifiable surface, nanoparticles have unique advantages compared with other drug carriers. As a research focus in recent years, polyethylene glycol-polylactic acid (PEG-PLA) block copolymer and its end-group derivative nanoparticles can enhance
R Gref et al.
Science (New York, N.Y.), 263(5153), 1600-1603 (1994-03-18)
Injectable nanoparticulate carriers have important potential applications such as site-specific drug delivery or medical imaging. Conventional carriers, however, cannot generally be used because they are eliminated by the reticulo-endothelial system within seconds or minutes after intravenous injection. To address these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service