916307
Platinum nanospheres
citrate functionalized, 30 nm diameter, 0.05 mg/mL (2 mM aqueous sodium citrate buffer)
Synonym(s):
Platinum nanoparticles, PtNP
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.23
Recommended Products
form
nanospheres
Quality Level
concentration
0.05 mg/mL (2 mM aqueous sodium citrate buffer)
diameter
30 nm
functional group
citrate
storage temp.
2-8°C
SMILES string
[Pt]
InChI
1S/Pt
InChI key
BASFCYQUMIYNBI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Platinum nanoparticles have many applications ranging from the automotive, chemical, and biomedical industries.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pt-based composite nanoparticles for magnetic, catalytic, and biomedical applications.
Liu Y, et al.
Journal of Materials Chemistry, 21, 12579-12587 (2011)
Deborah Pedone et al.
Chemical Society reviews, 46(16), 4951-4975 (2017-07-12)
Oxidative stress-dependent inflammatory diseases represent a major concern for the population's health worldwide. Biocompatible nanomaterials with enzymatic properties could play a crucial role in the treatment of such pathologies. In this respect, platinum nanoparticles (PtNPs) are promising candidates, showing remarkable
Recent Advances in FePt Nanoparticles for Biomedicine.
Shi Y, et al.
Journal of Nanomaterials, 2015, 1-13 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service