Skip to Content
Merck
All Photos(1)

Key Documents

915831

Sigma-Aldrich

Bioactive glass

powder, 13-93, ≥98%, 50-250 μm particle size

Synonym(s):

Bioglass ceramics, Silica glass

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaO:SiO2:P2O5:MgO:K2O:Na2O
UNSPSC Code:
12352305
NACRES:
NA.23

Pricing and availability is not currently available.

Quality Level

Assay

≥98%

form

powder

particle size

50-250 μm

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
915580915335915076
assay

≥98%

assay

≥98%

assay

≥98%

assay

≥98%

particle size

50-250 μm

particle size

50-250 μm

particle size

0.2-20 μm

particle size

0.2-20 μm

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

Application

Bioactive glass is able to form a hydroxyapatite-like surface layer thus providing a stable bond to bone cells. In addition, bioactive glass has been shown to stimulate bone cells regeneration. Due to its biocompatability and osteogenic capacity applications include tissue engineering, enamel reconstruction, bio-coatings, and high temperature adhesives.[1][2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Lungs, Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Baino et al.
Journal of functional biomaterials, 9(1) (2018-03-23)
Bioactive glasses caused a revolution in healthcare and paved the way for modern biomaterial-driven regenerative medicine. The first 45S5 glass composition, invented by Larry Hench fifty years ago, was able to bond to living bone and to stimulate osteogenesis through
Julian R Jones
Acta biomaterialia, 9(1), 4457-4486 (2012-08-28)
Bioactive glasses are reported to be able to stimulate more bone regeneration than other bioactive ceramics but they lag behind other bioactive ceramics in terms of commercial success. Bioactive glass has not yet reached its potential but research activity is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service