915394
Lithium Aluminum Titanium Phosphate (LATP) powder
battery grade
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Al0.3Li1.3Ti1.7(PO4)3
CAS Number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23
Recommended Products
grade
battery grade
Quality Level
Assay
≥99.5%
form
powder
mol wt
Mw 382.01 g/mol
composition
Li1.4Al0.4Ti1.6(PO4)3
surface area
0.86 m2/g
particle size
2-3 μm (average)
conductivity
0.01-0.1 mS/cm (ionic conductivity, cold-pressed pellet at room temperature)
density
2.92 g/cm3 (lit.)
application(s)
battery manufacturing
Application
Solid electrolytes (SEs) have become a practical option for lithium ion and lithium metal batteries because of their improved safety over commercially available ionic liquids. Solid electrolyte materials are highly conductive and made for advanced solid-state lithium ion battery technologies. These solutions are high purity and battery grade thus making them suitable as standards in LIB research.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lithium battery chemistries enabled by solid-state electrolytes.
Manthiram A, et al.
Nature Reviews. Materials, 2(4), 16103-16103 (2017)
Review on solid electrolytes for all-solid-state lithium-ion batteries
Zheng F, et al.
Journal of Power Sources, 389, 198-213 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service