Skip to Content
Merck
All Photos(1)

Key Documents

913952

Sigma-Aldrich

Titania nanoparticles

dispersion, 10 wt. % in ethanol, 700 nm particle size

Synonym(s):

TiO2 nanoparticles, TiOxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

form

dispersion
nanoparticles

Quality Level

concentration

10 wt. % in ethanol

particle size

700 nm

SMILES string

[Ti](=O)=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Titania nanoparticles are useful for a wide variety of applications, due to their high refractive index and large bandgap, providing optical properties in the visible spectrum. Applications of titania nanoparticles range from energy to medical applications.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modified titanium oxide (TiO2) nanocomposites and its array of applications: a review
Ghosh S S and Das A P
Toxicological & Environmental Chemistry, 97:5, 491-514 (2015)
Janusz Bogdan et al.
Nanoscale research letters, 12(1), 225-225 (2017-03-30)
Cancer has become a global problem. On all continents, a great number of people are diagnosed with this disease. In spite of the progress in medical care, cancer still ends fatal for a great number of the ill, either as
Mini review on photocatalysis of titanium dioxide nanoparticles and their solar applications.
Lan Y, et al.
Nano Energy, 2(5), 1031-1045 (2013)
Introduction: titanium dioxide (TiO2) nanomaterials
Chen X and Selloni A
Chemical Reviews, 114(19), 9281-9282 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service