Skip to Content
Merck
All Photos(2)

Key Documents

913758

Sigma-Aldrich

Phen-NaDPO

Synonym(s):

3-[6-(diphenylphosphinyl)-2-naphthalenyl]-1,10-Phenanthroline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H23N2OP
CAS Number:
Molecular Weight:
506.53
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥98%

Quality Level

form

powder

transition temp

Tg ~116 °C

solubility

soluble (Soluble in polar and weakly polar solvents such as isopropanol, toluene and xylenes)

semiconductor properties

(mobility=0.0001-0.001 cm2/V·s) (electron)

SMILES string

[P](=O)(c7ccccc7)(c6ccccc6)c1cc2c(cc(cc2)c3cnc4c5ncccc5ccc4c3)cc1

InChI

1S/C34H23N2OP/c37-38(30-9-3-1-4-10-30,31-11-5-2-6-12-31)32-18-17-25-20-26(14-15-27(25)22-32)29-21-28-16-13-24-8-7-19-35-33(24)34(28)36-23-29/h1-23H

InChI key

OAOXYBSWOZWDBB-UHFFFAOYSA-N

Application

Phen-NaDPO is a universal cathode modifier for organic optoelectronics. Phen-NaDPO can significantly lower the work function of the Ag metal as well as ITO and HOPG.
It lowers the energy loss between organic layer and the metal electrode, results in higher acheiveable efficiency for organic solar cells.[1] It is also used as cathode interfacial material for inverted Perovskite solar cells[2], and electron transport materials for Perovskite solar cells[3] and other organic electronic devices in general.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Triarylphosphine Oxide as Cathode Interfacial Material for Inverted Perovskite Solar Cells.
Wang K, et al.
Advanced Materials Interfaces, 6(12) (2019)
Use of the Phen-NaDPO:Sn(SCN)2 Blend as Electron Transport Layer Results to Consistent Efficiency Improvements in Organic and Hybrid Perovskite Solar Cells.
Seitkhan A, et al.
Advances in Functional Materials, 29(49), 1905810-1905810 (2019)
Qiang Su et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(10), 1800549-1800549 (2018-10-26)
The phenomenon of positive aging, i.e., efficiency increased with time, is observed in quantum-dot light-emitting diodes (QLEDs). For example, the external quantum efficiency (EQE) of blue QLEDs is significantly improved from 4.93% to 12.97% after storage for 8 d. The
Improved quantum dot light-emitting diodes with a cathode interfacial layer.
Ding T, et al.
Organic Electronics, 32, 89-93 (2016)
Tuning of the conformation of asymmetric nonfullerene acceptors for efficient organic solar cells.
Yang L,et al.
Journal of Material Chemistry A, 7(39), 22279-22286 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service