Skip to Content
Merck
All Photos(1)

Key Documents

912522

Sigma-Aldrich

Adamantanyl 2,2-diiodoacetate

≥95%

Synonym(s):

Suero cyclopropanation reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16I2O2
CAS Number:
Molecular Weight:
446.06
UNSPSC Code:
12352200

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: C-C Bond Formation

mp

94-95 °C

storage temp.

2-8°C

Application

Adamantanyl 2,2-diiodoacetate is a photocyclopropanation reagent capable of late-stage functionalization. The cyclopropanation of styrenes proceeds in the presence of visible light, without the need of a photocatalyst.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Legal Information

This product is not available in Europe.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A transition-metal-free & diazo-free styrene cyclopropanation
Herraiz A G and Suero M G
Chemical Science, 10, 9374-9379 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service