912239
Cerium Zirconium oxide nanofiber (Ce:Zr ratio 50:50)
Synonym(s):
CeZrO4 nanofiber, Nanofibrous inorganic powder structure
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
CeZrO4
CAS Number:
Molecular Weight:
295.34
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
Recommended Products
form
nanofiber
Quality Level
fiber diam.
300-800 nm × 2-hundreds of μm , structure: Polycystalline nanofibers
surface area
30-60 m2/g
primary crystallite size
8 nm (Crystal Phase: tetragonal)
SMILES string
[Ce](=O)=O.[Zr](=O)=O
InChI
1S/Ce.4O.Zr
InChI key
WKROXIWGJAIPLZ-UHFFFAOYSA-N
Application
Cerium Zirconium oxide has applications in automotive combustion sensor and controller for catalytic convertors, catalyst support for hot gas desulfurization, fuel cells, catalyst and catalyst support, thermochemical water splitting, infrared filters, and inorganic separators.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service