911860
NanoFabTx™ microfluidic - micro
device kit for synthesis of 1-5 μm particles
Synonym(s):
Microfluidic kit, Microparticle, NanoFabTx™ device kit, Nanoformulation
About This Item
Recommended Products
description
Microfludic device kit for synthesizing 1-5 μm microparticles
Kit components :
Quality Level
application(s)
advanced drug delivery
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Step-by-step microfluidic-based protocols developed and tested by our formulation scientists
- Optimized to make monodisperse microparticles of any size between 1 and 5 μm
- Facilitates simple optimization of drug encapsulation and loading efficiency in microparticles
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service