Skip to Content
Merck
All Photos(1)

Key Documents

910821

Sigma-Aldrich

Titanium aluminium carbide 211

MAX Phase, ≥80%, ≤100 μm particle size

Synonym(s):

MAX Phase 211, Ti2AlC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti2AlC
CAS Number:
NACRES:
NA.23

Assay

≥80%

form

powder

color

dark gray

particle size

≤100 μm

Looking for similar products? Visit Product Comparison Guide

General description

Mxenes find extensive use in LiBs and rechargeable non-lithium-ion (Na+, K+, Mg2+, Ca2+, and Al3+) batteries . This MXene precursor enables the potential of Ti3C2Tx Mxenes as anode , cathode or separator to boost battery life-cycle and efficiency.

Application

MAX phases are a family of ternary carbides and nitrides that share a similar layered hexagonal crystal structure.
Ti2AlC MAX phase exhibits high-temperature stability, thermal shock resistance, damage tolerance, crack-healing capability, good machinability, and exceptional oxidation resistance (immune to thermal cycling), and was widely used for high-temperature applications such as high-temperature heating elements, gas burner nozzles and industrial die inserts.

MAX phases are important precursors for synthesizing MXene, a highly conductive 2-dimentional nanomaterial. MXenes are produced by selective etching of the A element from the MAX phases. It combine the metallic conductivity of transition metal carbides with the hydrophilic nature of their hydroxyl or oxygen terminated surfaces. Ti2AlC MAX phase is one of the most used MAX phase for MXene (Ti2CTx).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MXenes for advanced separator in rechargeable batteries
An, Yongling et. al.
Materials Today, 57, 146-179 (2022)
Prediction and Characterization of MXene Nanosheet Anodes for Non-Lithium-Ion Batteries
Xie, Yu et. al.
ACS Nano, 8, 9606?9615- 9606?9615 (2014)
MAX phases: bridging the gap between metals and ceramics.
Radovic M ,et al.
American Ceramic Society Bulletin, 92(3), 20-27 (2013)
2D metal carbides and nitrides (MXenes) for energy storage.
Anasori B, et al.
Nature Reviews. Materials, 2(2), 16098-16098 (2017)
MXene as a novel intercalation-type pseudocapacitive cathode and anode for capacitive deionization
Srimuk,Pattarachai
Journal of Materials Chemistry, 4, 18265-18271 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service