Skip to Content
Merck
All Photos(1)

Key Documents

907278

Sigma-Aldrich

H-L-Photo-leucine HCl

≥98%

Synonym(s):

(S)-2-Amino-3-(3-methyl-3H-diazirin-3-yl)propanoic acid hydrochloride, (S)-2-Amino-3-(3H-diazirin-3-yl)butanoic acid hydrochloride, Diazirine amino acid, Photo-Leu, Photo-crosslinking amino acid, Photoprobe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10ClN3O2
Molecular Weight:
179.60
UNSPSC Code:
12352209

Assay

≥98%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

availability

available only in USA

application(s)

peptide synthesis

storage temp.

−20°C

Application

H-L-Photo-leucine HCl is a diazirine-containing leucine amino acid and multifunctional photo-crosslinker. Its incorporation into peptides or small-molecule probes and tools allows for photoaffinity labeling of cellular targets and protein-protein interactions upon UV light (∼360 nm) irradiation to form a covalent bond. This and other multifunctional probe building blocks will continue to accelerate drug discovery research for probing cellular mechanisms, target ID/validation, and understanding traditionally undruggable targets. An Fmoc-protected version is also available as 907391.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yuan Gao et al.
Chemical communications (Cambridge, England), 48(67), 8404-8406 (2012-07-18)
Here we report the first example of the use of supramolecular hydrogels to discover the protein targets of aggregates of small molecules.
Balakrishnan S Moorthy et al.
Journal of visualized experiments : JoVE, (98), 52503-52503 (2015-05-06)
Amide hydrogen/deuterium exchange (ssHDX-MS) and side-chain photolytic labeling (ssPL-MS) followed by mass spectrometric analysis can be valuable for characterizing lyophilized formulations of protein therapeutics. Labeling followed by suitable proteolytic digestion allows the protein structure and interactions to be mapped with
Christopher J Shaffer et al.
Journal of the American Society for Mass Spectrometry, 27(4), 633-645 (2016-01-29)
Noncovalent complexes of hydrophobic peptides GLLLG and GLLLK with photoleucine (L*) tagged peptides G(L* n L m )K (n = 1,3, m = 2,0) were generated as singly charged ions in the gas phase and probed by photodissociation at 355
Bojie Zhang et al.
Journal of the American Society for Mass Spectrometry, 27(3), 552-555 (2015-12-19)
Protein footprinting combined with mass spectrometry provides a method to study protein structures and interactions. To improve further current protein footprinting methods, we adapted the fast photochemical oxidation of proteins (FPOP) platform to utilize carbenes as the footprinting reagent. A
Christopher J Shaffer et al.
Journal of the American Society for Mass Spectrometry, 27(7), 1176-1185 (2016-04-10)
We report a combined experimental and computational study aimed at elucidating the structure of N-terminal fragment ions of the c type produced by electron transfer dissociation of photo-leucine (L*) peptide ions GL*GGKX. The c 4 ion from GL*GGK is found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service