Skip to Content
Merck
All Photos(1)

Key Documents

905836

Sigma-Aldrich

Gold nanospheres

100 nm, FITC and protein A functionalized, powder

Synonym(s):

Dye labeled gold nanoparticles, FITC labeled gold nanospheres, FITC tagged gold nanospheres, Fluorescein isothiocyanate labeled gold nanospheres, Fluorescent labeled gold nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

nanospheres
powder

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Gold nanospheres (AuNS) (100nm, FITC and protein A functionalized, powder) are gold nanoparticles with unique properties which include biocompatibility, easy surface modification, and excellent photothermal conversion. They can be synthesized by a variety of methods such as galvanic replacement of gold nanoparticles, templated galvanic replacement of silver for gold, and electrochemical methods.

Application

Owing to size and shape dependent quantum effects, the gold nanoparticles exhibit unique surface plasmon resonance absorption, scattering, fluorescence, and photothermal properties which render them suitable for applications in catalysis, chemical sensing, biosensing , cellular and bioimaging, drug and gene delivery, and photothermal therapy. The fluorescent labeling of gold nanoparticles have been shown to cause emission enhancement in fluorophores. This makes the fluorescence tagged gold nanoparticles a dual mode nanoprobe for drug delivery and bioimaging applications.

Legal Information

Product of Nanopartz Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescently Labeled Gold Nanoparticles with Minimal Fluorescence Quenching
Y Lu, et al.
The Journal of Physical Chemistry C, 114, 17446-17446 (2010)
Preparation, modification, and application of hollow gold nanospheres
Ren Q, et al.
Journal of Nanomaterials, 2015 (2015)
Gold Nanoparticles for In Vitro Diagnostics.
Wen Zhou et al.
Chemical reviews, 115(19), 10575-10636 (2015-06-27)
Xiaohua Huang et al.
Advanced materials (Deerfield Beach, Fla.), 21(48), 4880-4910 (2009-12-28)
Noble metal nanoparticles are capable of confining resonant photons in such a manner as to induce coherent surface plasmon oscillation of their conduction band electrons, a phenomenon leading to two important properties. Firstly, the confinement of the photon to the
Parwathy Chandran et al.
Nanotoxicology, 11(4), 507-519 (2017-04-20)
This study investigated the role of nanoparticle size and surface chemistry on biocorona composition and its effect on uptake, toxicity and cellular responses in human umbilical vein endothelial cells (HUVEC), employing 40 and 80 nm gold nanoparticles (AuNP) with branched polyethyleneimine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service