Skip to Content
Merck
All Photos(2)

Key Documents

901934

Sigma-Aldrich

NORIT® RX3 EXTRA

Synonym(s):

Activated carbon, Activated charcoal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C
CAS Number:
MDL number:
UNSPSC Code:
23201100
NACRES:
NA.23

description

Apparent Density: 380 k/m3
Ash content: 2 mass%
Butane adsorption at p/p0=0.1 : 28g/100g

form

powder

hardness

99 (Ball-pan hardness)

anion traces

chloride (Cl-): 0.1 mass% (acid extr.)

cation traces

Cu: 10 mg/kg (acid extr.)
Zn: 2 mg/kg (acid extr.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Application

  • Activated carbons are internally porous microcrystalline, non-graphitic forms of carbon. The activated carbons possess large surface area (~1000 m2/g) and pore volumes which make them useful for applications in energy storage devices, catalysis, and for removal of impurities from gases and liquids.
  • NORIT® RX 3 EXTRA is an acid washed, steam activated carbon that exhibits high pore volume. Its excellent purity along with good abrasion resistance makes it suitable as a catalyst or catalyst carrier for gas phase applications requiring high purity activated carbon.
NORIT® RX3 EXTRA (AC) can be used in a variety of applications such as solar collector, removal of metal from wastewater and production of hydrogen.

Legal Information

Product of Cabot Corporation.
NORIT® is a registered trademark of Cabot Corporation.
Norit is a registered trademark of Norit N.V.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of iron-impregnated granular activated carbon for arsenic removal from drinking water
Chang Q, et al.
Journal of Hazardous Materials, 184(1-3), 515-522 (2010)
Fundamentals of Adsorption onto Activated Carbon in Water and Wastewater Treatment.
Aktas O, et al.
Wiley-VCH (2011)
Improving ice productivity and performance for an activated carbon/methanol solar adsorption ice-maker
Qasem NAA and El-Shaarawi MAI
Solar Energy, 98, 523-542 (2013)
Hydrogen production by propylene-assisted decomposition of methane over activated carbon catalysts
Malaika A and Kozlowski M
International Journal of Hydrogen Energy, 35(19), 10302-10310 (2010)
Energy storage applications of activated carbons: supercapacitors and hydrogen storage.
Sevilla M and Mokaya R
Energy & Environmental Science, 7, 1250-1280 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service