Skip to Content
Merck
All Photos(3)

Documents

901907

Sigma-Aldrich

RuPhos

95%

Synonym(s):

2-Dicyclohexylphosphino-2′,6′-diisopropoxybiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H43O2P
CAS Number:
Molecular Weight:
466.64
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.22

Assay

95%

form

powder or crystals

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

reagent type: ligand

mp

123-126 °C
125 °C

functional group

phosphine

SMILES string

CC(C)Oc1cccc(OC(C)C)c1-c2ccccc2P(C3CCCCC3)C4CCCCC4

InChI

1S/C30H43O2P/c1-22(2)31-27-19-13-20-28(32-23(3)4)30(27)26-18-11-12-21-29(26)33(24-14-7-5-8-15-24)25-16-9-6-10-17-25/h11-13,18-25H,5-10,14-17H2,1-4H3

InChI key

MXFYYFVVIIWKFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bulky phosphine ligand used in a palladium-catalyzed cross-coupling of aminoethyltrifluoroborates with electron-poor aryl bromides.

Legal Information

Usage subject to Patents: EP 1097158; JP 5758844; CA 2336691

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Chatelain et al.
Angewandte Chemie (International ed. in English), 58(42), 14959-14963 (2019-08-24)
Ideal organic syntheses involve the rapid construction of C-C bonds, with minimal use of functional group interconversions. The Suzuki-Miyaura cross-coupling (SMC) is a powerful way to form biaryl linkages, but the relatively similar reactivity of electrophilic partners makes iterative syntheses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service