Skip to Content
Merck
All Photos(1)

Key Documents

901599

Sigma-Aldrich

Carbon disulfide solution

5 M in ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

concentration

5 M in ethanol

refractive index

n/D 1.387

density

0.91613 g/mL

SMILES string

S=C=S

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Valuable building block offered as a solution in ethanol for more convenient handling

related product

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Target Organs

Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service