900919
Azidoacetic acid NHS ester
Synonym(s):
(2,5-Dioxopyrrolidin-1-yl) 2-azidoacetate, 1-[(Azidoacetyl)oxy]pyrrolidine-2,5-dione
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H6N4O4
CAS Number:
Molecular Weight:
198.14
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22
Recommended Products
form
powder or crystals
Quality Level
reaction suitability
reagent type: cross-linking reagent
availability
available only in USA
storage temp.
−20°C
SMILES string
[N+](=[N-])=NCC(=O)ON1C(=O)CCC1=O
InChI
1S/C6H6N4O4/c7-9-8-3-6(13)14-10-4(11)1-2-5(10)12/h1-3H2
InChI key
FENNDBOWHRZLTQ-UHFFFAOYSA-N
Application
Azidoacetic acid NHS ester is a simple azido-containing reagent with an N-hydroxysuccinimide (NHS) ester that can be reacted with primary amines at pH 7-9 forming a stable amide bond. The azide allows for chemoselective ligation with an alkyne or cyclooctyne. It is generally used to functionalize amines and amino acids with azidoacetyl groups. It is generally used in click chemistry applications.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Self-react. C
Storage Class Code
5.2 - Organic peroxides and self-reacting hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Click and Pick: Identification of Sialoside Analogues for Siglec?Based Cell Targeting.
Rillahan C D, et al.
Angewandte Chemie (International Edition in English), 51(44), 11014-11018 (2012)
Tension sensing nanoparticles for mechano-imaging at the living/nonliving interface.
Liu Y, et al.
Journal of the American Chemical Society, 135(14), 5320-5323 (2013)
Synthetic glycosphingolipids for live-cell labeling.
Dauner M, et al.
Bioconjugate Chemistry, 27(7), 1624-1637 (2016)
Nanoparticle tension probes patterned at the nanoscale: impact of integrin clustering on force transmission.
Liu Y, et al.
Nano Letters, 14(10), 5539-5546 (2014)
In vivo targeting of hydrogen peroxide by activatable cell-penetrating peptides.
Weinstain R, et al.
Journal of the American Chemical Society, 136(3), 874-877 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service