900757
Sodium pyridine-2-sulfinate
Synonym(s):
2-Pyridinesulfinic acid, Sodium salt, Willis 2-pyridylsulfinates H
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
form
powder or crystals
Quality Level
reaction suitability
reaction type: C-C Bond Formation
mp
292 °C
functional group
sulfinic acid
SMILES string
[Na+].[S](=O)([O-])c1ncccc1
InChI
1S/C5H5NO2S.Na/c7-9(8)5-3-1-2-4-6-5;/h1-4H,(H,7,8);/q;+1/p-1
InChI key
HQCGASCBCQJRDJ-UHFFFAOYSA-M
Application
This 2-pyridylsulfinate enables Suzuki-Miyaura cross coupling with aryl halides to access diverse pyridine derivatives as reported by the laboratory of Michael Willis.. No longer impeded by unstable 2-pyridyl boronates, the sulfinate alternatives provide Pd-catalyzed coupling routes to therapeutically valuable pyridine rings.
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service