Skip to Content
Merck
All Photos(1)

Key Documents

900560

Sigma-Aldrich

Graphene quantum dots

>0.005% in H2O

Synonym(s):

GQDs

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101522
NACRES:
NA.23

form

liquid

Quality Level

concentration

>0.005% in H2O

fluorescence

, quantum yield >50%

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Graphene quantum dots (GQDs) have the potential to be applied towards next generation electronics and energy applications. Graphene itself does not possess band gap energy and its usage is limited. However, when their size becomes as small as quantum dots, they exhibit photoluminescence in the ultraviolet, visible, and near infrared ranges. The size of graphene quantum dots ranges approximately 1-4 nm. Due to their quantum confinement effect, GQDs can be applied to photovoltaic devices such as solar cells, bio imaging (cancer cell imaging, protein analysis, cell tracking), various type of sensors, etc.

Application

  • Bio-imaging.
  • Photocatalysis.
  • LED.
  • Energy conversion.
  • Sensing.

Storage Class Code

10 - Combustible liquids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service