Skip to Content
Merck
All Photos(2)

Key Documents

900349

Sigma-Aldrich

cataCXium A Pd G4

Synonym(s):

[di(adamantan-1-yl)(butyl)phosphine](methanesulfonato-κO)[2′-(methylamino)-2-biphenylyl]palladium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H54NO3PPdS
CAS Number:
Molecular Weight:
742.30
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

powder

Quality Level

feature

generation 4

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

SMILES string

CCCCP([C@@]12C[C@H]3C[C@@H](C1)C[C@@H](C2)C3)[C@@]4(C5)C[C@H]6C[C@H](C[C@@H]5C6)C4.CNC7=C(C8=CC=CC=C8[Pd]OS(C)(=O)=O)C=CC=C7

InChI

1S/C24H39P.C13H12N.CH4O3S.Pd/c1-2-3-4-25(23-11-17-5-18(12-23)7-19(6-17)13-23)24-14-20-8-21(15-24)10-22(9-20)16-24;1-14-13-10-6-5-9-12(13)11-7-3-2-4-8-11;1-5(2,3)4;/h17-22H,2-16H2,1H3;2-7,9-10,14H,1H3;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

ROZFJYIWIVSDBR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

A fourth generation (G4) Buchwald precatalyst that is similar to the third generation (G3) precatalysts except that the amino group on the biphenyl backbone is methylated. This modification helps to prevent the limitations found when using the third generation precatalysts. It is air, moisture, and thermally-stable and shows good solubility in common organic solvents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Rupture of the pregnant uterus].
I Pardini et al.
Rivista italiana di ginecologia, 58(6), 481-502 (1977-11-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service