900172
Fluorescent nanodiamond
Nitrogen vacancy ≤4 NV/particle, 35 nm avg. part. size (DLS), 1 mg/mL in deionized water, carboxylated
Synonym(s):
Nanodiamond
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352302
NACRES:
NA.23
Recommended Products
description
Zeta potential +65.1 mV ±20%
Quality Level
form
dispersion
nanoparticles
composition
Nitrogen vacancy, ≤4 NV/particle
concentration
1 mg/mL in deionized water
avg. part. size
35 nm (DLS)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Nanodiamonds containing nitrogen-vacancy (NV) centers can be prepared using irradiation with high energy electrons. Nanodiamonds with low numbers of NV centers (ideally, single defect centers) are suitable for quantum signaling, optics, and computing applications because their fluorescence allows for observation of single quantum systems. These particles have approximately 1-4 NV centers per particle as measured using AFM confocal microscopy with subsequent derivation of the correlation function using a Hanbury-Brown-Twiss setup.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Scanning Confocal Optical Microscopy and Magnetic Resonance on Single Defect Centers.
Gruber A. et al.
Science, 276, 2012-2014 (1997)
High yield fabrication of fluorescent nanodiamonds.
Boudou, J.P, et al.
Nanotechnology, 20 (2009)
In Commercial quantities of ultrasmall fluorescent nanodiamonds containing color centers.
Shenderova O, et al.
Advances in Photonics of Quantum Computing, Memory, and Communication X, San Francisco, SPIE: San Francisco (2017)
Single defect centres in diamond: A review.
Jelezko F, et al.
Physica Status Solidi, 203(13), 3207-3225 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service