Skip to Content
Merck
All Photos(1)

Key Documents

857858

Sigma-Aldrich

5-Iodouracil

98%

Synonym(s):

2,4-Dihydroxy-5-iodopyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3IN2O2
CAS Number:
Molecular Weight:
237.98
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

274-276 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

IC1=CNC(=O)NC1=O

InChI

1S/C4H3IN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)

InChI key

KSNXJLQDQOIRIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J D Robertson et al.
Pharmaceutical research, 9(11), 1410-1414 (1992-11-01)
X-ray fluorescence (XRF) has been used to determine in vivo the percutaneous absorption of 5-iodouracil (5IU) in dimethyl sulfoxide (DMSO) on female Sprague-Dawley rats. An average absorption rate constant of 122 +/- 34 micrograms/cm2-hr was obtained from the XRF measurements
N Belotto et al.
Journal of chromatography, 572(1-2), 327-332 (1991-12-06)
A new assay is described for 2'-deoxy-5-iodouridine, a drug employed as an antiviral agent by topical application. The parent drug, its systemic metabolite 5-iodouracil and an internal standard (5-iodouridine) were extracted from salted serum by an ethyl acetate partition at
Takanori Oyoshi et al.
Journal of the American Chemical Society, 124(10), 2086-2087 (2002-03-07)
Photoirradiation of 5-iodouracil-containing DNA, d(GTAAT(I)UAC)(2) with Sso7d protein, possessing significant kink in DNA in the crystal structure induces an unprecedented intrastrand H abstraction at the methyl group of T(5), together with selective photooxidations at Met29 of Sso7d. The reactivity of
S Sastry et al.
Biochemistry, 38(16), 4972-4981 (1999-04-23)
Transcription is the fundamental process by which RNA is synthesized by RNA polymerases on double-stranded DNA templates. One structurally simple RNA polymerase is encoded by bacteriophage T7. T7 RNA polymerase is an excellent candidate for studying structural aspects of transcription
Xiaorong Sun et al.
Oncotarget, 6(33), 34732-34744 (2015-09-30)
Bortezomib, a novel proteasome inhibitor, has been approved for treating multiple myeloma and mantle cell lymphoma and studied pre-clinically and clinically for solid tumors. Preferential cytotoxicity of bortezomib was found toward hypoxic tumor cells and endothelial cells in vitro. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service