Skip to Content
Merck
All Photos(1)

Documents

83845

Sigma-Aldrich

N-(Cyanomethyl)acetamide

≥97.0% (elemental analysis)

Synonym(s):

2-(Acetamido)acetonitrile, AAN, Aceturic acid nitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHCH2CN
CAS Number:
Molecular Weight:
98.10
Beilstein:
1700811
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (elemental analysis)

form

solid

mp

73-80 °C

SMILES string

CC(=O)NCC#N

InChI

1S/C4H6N2O/c1-4(7)6-3-2-5/h3H2,1H3,(H,6,7)

InChI key

LHWCUMYXUCPQSZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mattias Strand et al.
The International journal of eating disorders, 53(10), 1729-1738 (2020-08-01)
Individuals meeting all criteria for anorexia nervosa (AN) except that weight falls within or above the normal range despite significant weight loss are categorized as having atypical AN (AAN). Existing research has provided mixed evidence concerning the diagnostic demarcation of
Ling-Yu Pang et al.
BMC neurology, 17(1), 164-164 (2017-08-27)
Acute autonomic neuropathy (AAN) is rare disorder with anecdotal report, especially for childhood onset patients. Misdiagnosis or delays in treatment can always be found in clinical practice. We conducted this study to give a description of the manifestations and treatment
K Jean Forney et al.
The International journal of eating disorders, 50(8), 952-962 (2017-04-25)
Atypical anorexia nervosa (AAN) is defined by the symptoms of anorexia nervosa in the presence of "significant weight loss" in individuals who are not underweight. Description of current weight in AAN has been limited, significant weight loss has not been
Jason M Nagata et al.
The International journal of eating disorders, 52(5), 591-596 (2019-02-17)
To compare bone mineral density (BMD) and body composition among adolescents: (a) with atypical anorexia nervosa (AAN) versus anorexia nervosa (AN) and (b) those with and without a prior history of overweight. Electronic medical records of patients 9-20 years with AN
Romergryko G Geocadin et al.
Neurology, 88(22), 2141-2149 (2017-05-12)
To assess the evidence and make evidence-based recommendations for acute interventions to reduce brain injury in adult patients who are comatose after successful cardiopulmonary resuscitation. Published literature from 1966 to August 29, 2016, was reviewed with evidence-based classification of relevant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service