Skip to Content
Merck
All Photos(1)

Key Documents

806617

Sigma-Aldrich

Silver

Nanowires, diam. × L 20 nm (±2 nm) × 12 μm (±2 μm), 5 mg/mL (in ETH)

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12141740
NACRES:
NA.23
Pricing and availability is not currently available.

description

Flash point 57.2 °F

Quality Level

form

suspension

concentration

5 mg/mL (in ETH)

diam. × L

20 nm (±2 nm) × 12 μm (±2 μm)

color

gray

density

0.7937 g/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Owing to unique physical and chemical properties the nanowires of silver find wide range of applications extending from catalysis to photovoltaics, touch screen display, biosensing etc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service