Skip to Content
Merck
All Photos(5)

Documents

806579

Sigma-Aldrich

Reduced graphene oxide

greener alternative

tetraethylene pentamine functionalized

Synonym(s):

Reduced graphene oxide-tetraethylene pentamine, rGO-TEPA, rGO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CxOyHz
UNSPSC Code:
12352103
NACRES:
NA.23

Quality Level

form

powder

composition

Carbon, >65 wt. %
Nitrogen, >8 wt. %

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

CN(CCNCCN)CCNCCN.[rGO]

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Click here for more information.

Application

Immobilization of antibodies for use in ultrasensitive electrochemical immunosensors using the abundant amine groups and a large electroconductive surface area of reduced graphene oxide-tetraethylene pentamine (rGO-TEPA).
Amount of NH2 groups: In order to estimate the amount of NH2 groups in rGO-TEPA a reaction with tetrabromophthalic anhydride was performed and the Br amount was quantified by the Schöniger flask test. The value obtained corresponds to 1.1 mmol NH2/g.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guolin Yuan et al.
Biosensors & bioelectronics, 69, 95-99 (2015-02-24)
In this work, a novel electrochemical immunosensor for the detection of secretoneurin (SN), which uses metal ion functionalised reduced graphene oxide-tetraethylene pentamine (rGO-TEPA) as a label, is reported for the first time. rGO-TEPA contains a large number of amino groups
Aiping Guo et al.
Biosensors & bioelectronics, 63, 39-46 (2014-07-25)
A novel and sensitive electrochemical immunosensor for ultrasensitive detection of pancreatic cancer biomarker carbohydrate antigen 199 (CA199) was proposed by using Au@Cu(x)OS yolk-shell nanostructures with porous shells as labels for signal amplification. Au@Cu(x)OS yolk-shell nanostructures exhibit high electrocatalytic activity toward
Dan Wu et al.
Biosensors & bioelectronics, 54, 634-639 (2013-12-18)
A novel, highly sensitive electrochemical immunoassay was proposed for the simultaneous determination of carcinoembryonic antigen (CEA) and squamous cell carcinoma antigen (SCCA) for the diagnosis of cervical cancer. Using an electrochemical analysis technique, two well-separated peaks were generated by neutral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service