806129
bis-MPA-Azide dendrimer
trimethylol propane core, generation 3
Synonym(s):
G3-TMP-Azide
About This Item
Quality Level
Assay
95%
form
liquid
storage temp.
−20°C
SMILES string
O=C(OCC(COC(C(COC(C(COC(C(COC(CCCCCN=[N+]=[N-])=O)(COC(CCCCCN=[N+]=[N-])=O)C)=O)(COC(C(COC(CCCCCN=[N+]=[N-])=O)(COC(CCCCCN=[N+]=[N-])=O)C)=O)C)=O)(COC(C(COC(C(COC(CCCCCN=[N+]=[N-])=O)(COC(CCCCCN=[N+]=[N-])=O)C)=O)(COC(C(COC(CCCCCN=[N+]=[N-])=O)(COC(CCCCCN
InChI
1S/C255H398N72O90/c1-23-255(186-415-231(370)252(20,180-409-225(364)246(14,168-397-213(352)234(2,144-373-189(328)96-48-24-72-120-280-304-256)145-374-190(329)97-49-25-73-121-281-305-257)169-398-214(353)235(3,146-375-191(330)98-50-26-74-122-282-306-258)147-376-192(331)99-51-27-75-123-283-307-259)181-410-226(365)247(15,170-399-215(354)236(4,148-377-193(332)100-52-28-76-124-284-308-260)149-378-194(333)101-53-29-77-125-285-309-261)171-400-216(355)237(5,150-379-195(334)102-54-30-78-126-286-310-262)151-380-196(335)103-55-31-79-127-287-311-263,187-416-232(371)253(21,182-411-227(366)248(16,172-401-217(356)238(6,152-381-197(336)104-56-32-80-128-288-312-264)153-382-198(337)105-57-33-81-129-289-313-265)173-402-218(357)239(7,154-383-199(338)106-58-34-82-130-290-314-266)155-384-200(339)107-59-35-83-131-291-315-267)183-412-228(367)249(17,174-403-219(358)240(8,156-385-201(340)108-60-36-84-132-292-316-268)157-386-202(341)109-61-37-85-133-293-317-269)175-404-220(359)241(9,158-387-203(342)110-62-38-86-134-294-318-270)159-388-204(343)111-63-39-87-135-295-319-271)188-417-233(372)254(22,184-413-229(368)250(18,176-405-221(360)242(10,160-389-205(344)112-64-40-88-136-296-320-272)161-390-206(345)113-65-41-89-137-297-321-273)177-406-222(361)243(11,162-391-207(346)114-66-42-90-138-298-322-274)163-392-208(347)115-67-43-91-139-299-323-275)185-414-230(369)251(19,178-407-223(362)244(12,164-393-209(348)116-68-44-92-140-300-324-276)165-394-210(349)117-69-45-93-141-301-325-277)179-408-224(363)245(13,166-395-211(350)118-70-46-94-142-302-326-278)167-396-212(351)119-71-47-95-143-303-327-279/h23-188H2,1-22H3
InChI key
MACMPYCWHGFIKL-UHFFFAOYSA-N
Related Categories
General description
- End Group Functionality: Azide surface group
- Generation: 3
- No. Surface Groups: 24
- Calculated Mol. Wt.: 5912.4 g/mol
- Other Notes: bis-MPA = 2,2-bis(hydroxymethyl)propionic acid
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service