Skip to Content
Merck
All Photos(1)

Key Documents

804568

Sigma-Aldrich

(R,S)-Bode Kinetic Resolution Catalyst

Synonym(s):

(4aR,9aS)-6-Bromo-4-hydroxy-4,4a,9,9a-tetrahydroindeno[2,1-b][1,4]oxazin-3(2H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10BrNO3
CAS Number:
Molecular Weight:
284.11
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

solution

Quality Level

functional group

bromo
ether

storage temp.

15-25°C

SMILES string

O=C(CO1)N(O)[C@@]2([H])[C@]1([H])CC3=CC=C(Br)C=C32

InChI

1S/C11H10BrNO3/c12-7-2-1-6-3-9-11(8(6)4-7)13(15)10(14)5-16-9/h1-2,4,9,11,15H,3,5H2/t9-,11+/m0/s1

InChI key

FQXSUQCCAHKURK-GXSJLCMTSA-N

Application

The following hydroxamic acid catalyst was shown by Bode and coworkers to assist in the kinetic resolution of cyclic amines through acylation of one enantiomer, which can be controlled using either the (R or S)- or (S or R)-Bode Kinetic Resolution Catalyst, both of which are also available through Sigma-Aldrich.
It can also be employed in the preparation of nonracemic piperidines by kinetic resolution using an α-hydroxy-α′, β-unsaturated ketone in the presence of a pyrrolotriazolium salt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concerted amidation of activated esters: Reaction path and origins of selectivity in the kinetic resolution of cyclic amines via N-heterocyclic carbenes and hydroxamic acid cocatalyzed acyl transfer
Allen SE, et al.
Journal of the American Chemical Society, 136(33), 11783-11791 (2014)
Catalytic kinetic resolution of disubstituted piperidines by enantioselective acylation: synthetic utility and mechanistic insights
Wanner B, et al.
Journal of the American Chemical Society, 137(35), 11491-11497 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service