796557
Stahl Aerobic Oxidation ABNO solution
0.04M ABNO in Acetonitrile, Solution for Oxidation of Primary and Secondary Alcohols
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352000
NACRES:
NA.22
Recommended Products
Quality Level
description
Freezing Point: 44.6°F
form
liquid
reaction suitability
reagent type: oxidant
refractive index
n/D 1.357
density
0.796 at 25 °C
storage temp.
2-8°C
Related Categories
General description
Stahl Aerobic Oxidation ABNO solution contains 1-methylimidazole (NMI), 2,2′-bipyridyl (bpy), and ABNO for the oxidation of primary and secondary alcohols.
Application
This convenience solution can be used in tandem with tetrakisacetonitrile copper(I) triflate (685038) for the oxidation of primary alcohols through aerobic conditions developed by the Stahl Research Group.
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
43.5 °F
Flash Point(C)
6.38 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Efficient Aerobic Oxidation of Secondary Alcohols at Ambient Temperature with an ABNO/NOx Catalyst System
Lauber, MB, and Stahl, SS.
ACS Catalysis, 3 (11), 2612?2616-2612?2616 (2013)
Janelle E Steves et al.
The Journal of organic chemistry, 80(21), 11184-11188 (2015-10-13)
Two solutions, one consisting of bpy/TEMPO/NMI and the other bpy/ABNO/NMI (bpy =2,2'-bipyridyl; TEMPO = 2,2,6,6-tetramethylpiperidine N-oxyl, ABNO = 9-azabicyclo[3.3.1]nonane N-oxyl; NMI = N-methylimidazole), in acetonitrile are shown to have good long-term stability (≥1 year) under air at 5 °C. The
Janelle E Steves et al.
Journal of the American Chemical Society, 135(42), 15742-15745 (2013-10-17)
Cu/TEMPO catalyst systems promote efficient aerobic oxidation of sterically unhindered primary alcohols and electronically activated substrates, but they show reduced reactivity with aliphatic and secondary alcohols. Here, we report a catalyst system, consisting of ((MeO)bpy)Cu(I)(OTf) and ABNO ((MeO)bpy = 4,4'-dimethoxy-2,2'-bipyridine;
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service