794279
Chloro(2-methylphenyl)bis(triphenylphosphine)nickel(II)
Synonym(s):
Buchwald Nickel Cross-Coupling Catalyst Precursor, [Bis(triphenylphosphine)](o-tolyl)chloronickel
About This Item
Recommended Products
form
solid
Quality Level
reaction suitability
core: nickel
reagent type: catalyst
mp
195-200 °C
storage temp.
2-8°C
SMILES string
Cl[Ni]C1=CC=CC=C1C.P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.P(C5=CC=CC=C5)(C6=CC=CC=C6)C7=CC=CC=C7
InChI
1S/2C18H15P.C7H7.ClH.Ni/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-7-5-3-2-4-6-7;;/h2*1-15H;2-5H,1H3;1H;/q;;;;+1/p-1
InChI key
QVGJKVLYWZGANE-UHFFFAOYSA-M
Related Categories
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 1B - Skin Sens. 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service